Chloracetamid ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
2-Chloroacetamide
C0086-25G
C0086-500G
TCIAC0086-25GEA
132
DKK
TCIAC0086-25G
TCIAC0086-500G
Chloracetamid ≥98.0% (by GC)
Chloracetamid
Formel:
C₂H₄ClNO Kogepunkt: 225°C Smeltepunkt: 115…117 °C Massefylde: 1,6 g/cm³ (20 °C) Flammepunkt: 170 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008027 CAS-nummer: 79-07-2 EINECS: 201-174-2 UN nummer: 2811 ADR: 6.1,III Merck Index: 14,02110 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystaline |
Purity (GC) | min. 98.0 % |
Melting point | 117.0 to 121.0 deg-C |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...