6-Nitroveratraldehyd ≥85.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4,5-Dimethoxy-2-nitrobenzaldehyde, 3,4-Dimethoxy-6-nitrobenzaldehyde

N0538-25G N0538-5G
TCIAN0538-25GEA 2960 DKK
TCIAN0538-25G TCIAN0538-5G
6-Nitroveratraldehyd ≥85.0% (by GC)
6-Nitroveratraldehyd
Formel: C₉H₉NO₅
Smeltepunkt: 134…135 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00007134
CAS-nummer: 20357-25-9
EINECS: 243-762-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1361893d048039447e522f4fd2501d72ba9efd95&vid=1361893d048039447e522f4fd2501d72ba9efd95&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 85.0 %
Melting point 132.0 to 136.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR