Bromacetonitril ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

B1058-25G B1058-500G B1058-100G
TCIAB1058-25GEA 509 DKK
TCIAB1058-25G TCIAB1058-500G TCIAB1058-100G
Bromacetonitril ≥97.0% (by GC)
Bromacetonitril
Formel: BrCNCH₂
Kogepunkt: 150°C
Smeltepunkt: –36 °C
Massefylde: 1,771 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001884
CAS-nummer: 590-17-0
EINECS: 209-672-1
UN nummer: 3276
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a886a303fdec4f98bb9a18ac3a37fe08e5533052&vid=a886a303fdec4f98bb9a18ac3a37fe08e5533052&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR