1-Brom-2-butin ≥96.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B2190-5G B2190-1G
TCIAB2190-5GEA 862 DKK
TCIAB2190-5G TCIAB2190-1G
1-Brom-2-butin ≥96.0% (by GC)
1-Bromo-2-butyne
Formel: C₄H₅Br
Flammepunkt: 36°C
MDL Number: MFCD00190233
CAS-nummer: 3355-28-0
UN nummer: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f4cb4c9e0af173b41f0c22b7abe90179b97912b&vid=2f4cb4c9e0af173b41f0c22b7abe90179b97912b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR