Kaliumtetrahydroborat ≥98.0% (V)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Potassium tetrahydridoborate, Potassium borohydride

P1681-100G P1681-25G
TCIAP1681-100GEA 591 DKK
TCIAP1681-100G TCIAP1681-25G
Kaliumtetrahydroborat ≥98.0% (V)
Potassium tetrahydroborate
Formel: H₄BK
Molekylvægt: 53.94 g/mol
Smeltepunkt: 500
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011396
CAS-nummer: 13762-51-1
EINECS: 237-360-5
UN nummer: 1870
ADR: 4.3 I
Merck Index: 14,07616
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=714f9a9fb6ac4954510dc2a61a307ac0e7893e04&vid=714f9a9fb6ac4954510dc2a61a307ac0e7893e04&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Gasburet method) min. 98.0 %
Purity (Sodium hypochlorite Method) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR