Unitec Laboratory Chairs
Leverandør: bimos
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9650-0551 9650-CI01 9650-2000 9650-3000 9650E-2571 9650E-9801 9651-0551 9651-CI01 9651-2000 9651-3000 9651E-2571 9651E-9801 9653-0551 9653-CI01 9653-2000 9653-3000 9653E-2571 9653E-9801 9653E-9802 9653-0562
BIMO9650-0551EA 2610 DKK
BIMO9650-0551 BIMO9650-CI01 BIMO9650-2000 BIMO9650-3000 BIMO9650E-2571 BIMO9650E-9801 132-1078 BIMO9651-CI01 BIMO9651-2000 BIMO9651-3000 132-1080 BIMO9651E-9801 132-1082 BIMO9653-CI01 BIMO9653-2000 BIMO9653-3000 132-1083 BIMO9653E-9801 BIMO9653E-9802 BIMO9653-0562
Unitec Laboratory Chairs
Stole
Ideal for the use in production and laboratory, this budget-priced, basic model has a generously proportioned seat and a high backrest. With black synthetic leather.
  • Washable, impervious to oil and disinfecting agents
Unitec Basic
Large, ergonomically designed backrest to take the strain off the spine and muscles with integrated lumbar support
Backrest height adjustment convenient and easy to handle in sitting position
Healthy posture due to a hump providing support for the back in the lumbar region
Ergonomically designed seat with rounded front edge, which prevents blocked circulation in the thighs
Stepless seat height setting supported by safety gas spring with protective cover
Permanent contact backrest: this mechanism ensures that the backrest responds immediately to any movements of the upper body by following them
Environment-friendly: the materials used are recyclable
5 black star base made from plastic for high stability, longetivity and safety; the flat design prevents stumbling hasard
Unitec ESD
The metal components have a conductive coating
The high-comfort upholstery is also conducive
With conductive glides or castors
Seat and backrest made from ESD fabric upholstery: resistant, breathable and comfortable
5 star base made from steel shape tube, conductive, powder-coated for high stability, longetivity and safety; the black flat design prevents stumbling hasard
Large, ergonomically designed backrest to take the strain off the spine and muscles with integrated lumbar support
Backrest height adjustment convenient and easy to handle in sitting position
Healthy posture due to a hump providing support for the back in the lumbar region
Ergonomically designed seat with rounded front edge, which prevents blocked circulation in the thighs
Stepless seat height setting supported by safety gas spring with protective cover
Permanent contact backrest: this mechanism ensures that the backrest responds immediately to any movements of the upper body by following them
Environment-friendly: the materials used are recyclable
Certificeringer: Complies with DIN 68 877 german GS 'safety tested' mark of confirmity
Confirms with quality management system DIN EN ISO 9001 standards
Complies with regulation EWG Nr. 1836/93 and environmental management system DIN ISO 14001 standards
Unitec 1,2,3 ESD: exceeds the requirements of ESD protection according to standard DIN EN 61340-5-1, 100 % ESD protection
Bestillingsoplysninger: The armrest and foot rings for the chairs can be ordered separately.