Industrial gloves, oil repellent, Hynit®
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
32-105/9 32-125/9 32-125/8 32-105/8 32-815/7.5 32-105/10 32-105/7.5 32-125/10 32-125/7 32-125/7.5 32-815/10 32-815/7 32-815/8 32-815/9 32-105/7
ANSE32-105/9EA 353 DKK
ANSE32-105/9 ANSE32-125/9 ANSE32-125/8 ANSE32-105/8 ANSE32-815/7.5 ANSE32-105/10 ANSE32-105/7.5 ANSE32-125/10 ANSE32-125/7 ANSE32-125/7.5 ANSE32-815/10 ANSE32-815/7 ANSE32-815/8 ANSE32-815/9 ANSE32-105/7
Industrial gloves, oil repellent, Hynit®
Handsker
These polyester/cotton liners are either fully nitrile impregnated or feature an impregnated palm. The oil repellent gloves provide mechanical protection.
  • Special nitrile formulation unites excellent resistance to abrasion with remarkable flexibility
  • Liquid-repellent impregnation repels oil, grease and dirt
  • Wide selection of styles, lengths and sizes allows you to choose the optimum glove for your particular application
Ideal for use in different applications, such as handling white goods, packaging, component assembly, shipping and receiving. Also suitable for use in steel mills and in the textile industry.
Certificeringer: EN 388:2016 (3111A), Cat. II