Scrubbers, K-415 series
Leverandør: BUCHI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
114152331 114152320 114152330 114152341
701-0238EA 60490 DKK
701-0238 701-0236 701-0237 701-0239
Scrubbers, K-415 series
Destillationssystemer Scrubber-Systemer
A modular, upgradeable, system for scrubbing of harmful fumes from distillation or digestion processes. The system can be obtained with up to four process stages depending on user requirements.
DuoScrub: Basic, two stage model with a neutralisation vessel and an absorption vessel linked to a powerful suction pump (140 W, up to 32 L/min.).
TripleScrub and TripleScrubECO: Three stage model with pre-condensation step as stage 1 to collect entrained liquids and avoid dilution of the neutralisation stage 2. The TripleScrubECO model is equipped with automatic cooling water control and connection to the digester to help optimise water usage.
QuadScrubECO: 4 stage model with condensation, neutralisation, absorption, and redox-reactions of specific fumes. The QuadScrubECO model is equipped with automatic cooling water control and connection to the digester to help optimise water usage.
4 stage model with condensation, neutralisation, absorption, and redox-reactions of specific fumes. The QuadScrubECO model is equipped with automatic cooling water control and connection to the digester to help optimise water usage.