Cut resistant gloves, ActivArmr® 28-329 / 28-359
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
28-359/10 28-359/9 28-359/8 28-329/10 28-329/8 28-329/9
ANSE28-359/10EA 175 DKK
ANSE28-359/10 ANSE28-359/9 ANSE28-359/8 ANSE28-329/10 ANSE28-329/8 ANSE28-329/9
Cut resistant gloves, ActivArmr® 28-329 / 28-359
Handsker
These protective gloves combine the outstanding protection of Kevlar®-reinforced construction with the grip and flexibility of Ansell’s advanced nitrile coating technology.
  • Lining is reinforced with Kevlar® fibres
  • NitraGold™ cuff for outstanding mechanical protection against cuts and abrasion
  • Antistatic according to EN 1149
  • Available with either nitrile palm or full coating
Ideal for safe handling of oily heavy metal parts. Suitable for steel bar manufacturing and handling, building materials handling, timber works and construction, sheet metal handling and metalwork.
Certificeringer: EN 388:2016 (4321C), EN 407 (X1XXXX); Cat. III