General purpose gloves, KSR®
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
22-515/7.5 22-515/8 22-515/10 22-515/7 22-515/9
ANSE22-515/7.5EA 854 DKK
ANSE22-515/7.5 ANSE22-515/8 ANSE22-515/10 ANSE22-515/7 ANSE22-515/9
General purpose gloves, KSR®
Handsker
This interlock cotton liner with full PVC coating is ideal for long-wearing light-duty work.
  • Light, flexible and comfortable: ideal for long wearing times
  • Excellent grip for handling small and difficult objects
  • Slip-on cuff style
Suitable applications include maintenance, shipping and receiving, product inspection, packaging, working with tools and handling aluminium profiles.
Certificeringer: EN 388:2016 (2121A); Cat. II