Racks for cryogenic vials, Nunc™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
331825 331826 331827 331828
479-1147EA 940 DKK
479-1147 479-1148 479-1149 479-1151
Racks for cryogenic vials, Nunc™
Stativer Stativer til Kryorør
  • Latch rack with standard ANSI microplate footprint
  • Holds 48 internally-threaded CryoTubes™ or Cryobank vials
  • Use pivoting lid on bench or lock it for transport and storage
  • Format facilitates automated picking/placing, capping/decapping, and multi-channel liquid handling
  • Designed for –80°C and liquid nitrogen (vapour phase) storage