Disposable spin columns for protein purification
Leverandør: G-Biosciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
786-725 786-727 786-724 786-726 786-720 786-719 786-718
786-725EA 1990 DKK
786-725 786-727 786-724 786-726 786-720 786-719 786-718
Disposable spin columns for protein purification
Filtre Centrifugefiltre
These disposable spin columns are designed for protein purification. These spin columns are uniquely designed, with a snap off end converting to a closure for the column for easy manipulation and use.
These columns can be spun with the stopper attached and then snapped off to elute solutions. For further storage or incubation, simply push the column into the stopper.