Safety spectacles, Adaptec™
Leverandør: HONEYWELL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1030749HS 1030759HS 1030739HS
DALL1030749HSEA 188 DKK
DALL1030749HS DALL1030759HS DALL1030739HS
Safety spectacles, Adaptec™
Briller
Stylish single lens safety glasses with vented, cushioned frame and PC lens.
  • Three position nose bridge is simple to adjust and keeps eyes centred in the lens
  • Self-adjusting nose pads
  • Adjustable lens angle reduces fogging; each temple hinge ratchets to seven positions
  • Temple microadjustment allows workers to adjust temple tension without removing eyewear
  • 4A+ coating on both sides of the lens ensures excellent anti-fog, anti-static and scratch resistant properties
Suitable for jobs where there are risks associated with abrasion, fogging, airborne particulates or impact.
Certificeringer: EN 166 FT