Safety spectacles, SP1000™
Leverandør: HONEYWELL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1028640 1028643 1028644
111-2161EA 233 DKK
111-2161 111-2162 111-2163
Safety spectacles, SP1000™
Briller
These dual lens spectacles provide protection against impact, wind, dust and airborne debris. Ideal for dirty, high particulate work environments such as grinding, chipping, shaving and sanding.
  • Soft and flexible nosepiece
  • Soft, high quality face cushioning and padded temple tips
  • Highly adjustable flame resistant headband
  • Wrap-around eight-base dual lens style
  • Top and bottom indirect ventilation and Dura-streme dual anti-fog/anti-scratch coating
Certificeringer: EN 166