Conductivity sensors
Leverandør: KNICK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ZU6985 SE204 SE202
663-2111EA 11360 DKK
663-2111 665-0297 665-0295
Conductivity sensors
Elektroder Elektroder til ledningsevnemåling
SE 202: 2-electrode sensor with integrated temperature probe (NTC 30 kohm) and flow cell.
SE 204: 4-electrode sensor with integrated temperature probe (NTC 30 kohm).
SE 615 MS: Digital 2-electrode sensor with integrated temperature probe (NTC 30 kohm) and Memosens connection (incl. adapter ZU0939).
ZU 6985: 4-electrode sensor with quick-reacting Pt 1000 temperature probe and KPG® tube.