Centrifuge tubes, cylindrical, short conical bottom
Leverandør: GLASWARENFABRIK KARL HECHT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
40946001 40947001 92094607001
525-1307EA 479 DKK
525-1307 HECH40947001 HECH92094607001
Centrifuge tubes, cylindrical, short conical bottom
Rør Centrifugerør
AR®-Glas, with or without graduations.
  • Rimless
  • Graduated tubes feature fused white 'ASSISTENT-PERMANENT' graduations