Centrifuge tubes, conical, with rim
Leverandør: GLASWARENFABRIK KARL HECHT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
40942002 40942001
525-0786EA 4230 DKK
525-0786 HECH40942001
Centrifuge tubes, conical, with rim
Rør Centrifugerør
AR®/soda glass or Duran® borosilicate glass 3,3.
  • Fused white graduation