Dual Gel Casters
Leverandør: HOEFER ELECTROPHORESIS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SE246 SE249 SE6009
HOEFSE246EA 460 DKK
HOEFSE246 HOEFSE249 HOEFSE6009
Dual Gel Casters
Blottingsæt
Includes casting cradle, casting clamp assemblies, sealing gasket set, and cams.
Bestillingsoplysninger: Glass plates, combs and spacers need to be ordered separately.