Cleanroom anti-fatigue mats, Ergomat® Hygiene
Leverandør: ERGOMAT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EH0304-RED EH0220-RED EH0220-GRN EH0304-BLU EH0220-GRA EH0304-GRN EH0209-GRN EH0306-BLU EH0306-GRA EH0306-GRN EH0306-RED EH0320-BLU EH0320-GRA EH0304-GRA EH0209-RED EH0406-BLU EH0209-GRA EH0209-BLU EH0206-RED EH0206-GRN EH0206-GRA EH0206-BLU EH0203-RED EH0203-GRN EH0203-GRA EH0203-BLU EH0220-BLU EH0320-GRN EH0320-RED EH0420-GRN EH0420-RED EH0420-GRA EH0420-BLU EH0412-RED EH0412-GRN EH0412-GRA EH0412-BLU EH0406-RED EH0406-GRN EH0406-GRA EH-RED0409 EH-GRE0204 EH0404-BLU EH0404-GRN EH0409-BLU EH0409-GRN EH0309-GRN EH0312-BLU EH0312-GRN EH0309-BLU EH0303-BLU EH0303-GRN EH0204-BLU EH0204-GRN EH0212-BLU EH0212-GRN EH-RED0404 EH-RED0312 EH-GRE0312 EH-GRE0309 EH-GRE0404 EH-GRE0409 EH-GRE0212 EH-GRE0303 EH-RED0212 EH-RED0309 EH-RED0303 EH-RED0204
115-0839EA 2510 DKK
115-0839 115-0838 115-0848 115-0859 115-0828 115-0849 115-0847 115-0860 115-0830 115-0850 115-0840 115-0861 115-0831 115-0829 115-0837 115-0862 115-0827 115-0857 115-0836 115-0846 115-0826 115-0856 115-0835 115-0845 115-0825 115-0855 115-0858 115-0851 115-0841 115-0854 115-0844 115-0834 115-0864 115-0843 115-0853 115-0833 115-0863 115-0842 115-0852 115-0832 115-1933 115-1920 115-1916 115-1917 115-1918 115-1919 115-1913 115-1914 115-1915 115-1912 115-1910 115-1911 115-1906 115-1907 115-1908 115-1909 115-1932 115-1931 115-1924 115-1923 115-1925 115-1926 115-1921 115-1922 115-1928 115-1930 115-1929 115-1927
Cleanroom anti-fatigue mats, Ergomat® Hygiene
Måtter Belastningsreducerende Måtter
Ergomat® Hygiene is made from pure nitrile rubber and is designed to increase comfort while reducing trip hazards, body pain, quality issues and sick leave. It is ideal for areas where heat, oil and petrochemical resistance is crucial.
  • Upper side with cones
  • Can be autoclaved (610×915 mm only) at 100 °C for up to 5 minutes
  • Frequent washes or chemical exposure will not affect the state of the mat; no curling up, swelling or expanding
  • Moulded, bevelled edges prevent curling, eliminating trip hazards
  • Raw, virgin material guarantee durability and ergonomic benefits
  • Free from silicone and latex
Suitable for use in Class ISO 5 (100) cleanrooms.
Colour: Blue, green, grey or red
Meets EN ISO/USDA/CFIA standards
Certificeringer: EN ISO/USDA/CFIA