Test tubes with or without rim, heavy wall
Leverandør: GLASWARENFABRIK KARL HECHT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
42788051 92278822450 42788081 42787024
HECH42788051EA 793 DKK
HECH42788051 HECH92278822450 HECH42788081 HECH42787024
Test tubes with or without rim, heavy wall
Rør Reagensglas
Thick-walled, made of borosilicate glass 3.3, exceptional mechanical strength.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes