Industrial knives, with exchangeable blades
Leverandør: SWANN MORTON
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4213 4212 4211 4214 4303 4302 4253 4252 4307
SWAN4213EA 337 DKK
SWAN4213 SWAN4212 SWAN4211 SWAN4214 SWAN4303 SWAN4302 SWAN4253 SWAN4252 SWAN4307
Industrial knives, with exchangeable blades
Knive
Heavy-duty cast metal handles with either fixed blade or retractable blade mechanism. Ideal for industrial applications as well as for D.I.Y. enthusiasts.
  • Fixed handle can be opened with the aid of a screwdriver or small coin for blade changing
  • Retractable handle can be opened by hand for blade changing
  • Handles delivered without blades, set includes blades