Spare seal rings for small flange connections
Leverandør: VACUUBRAND
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20660110 20660111 20660115 20660116 20660130 20660135 20660136 20660137
185-1331EA 41.7 DKK
185-1331 185-1333 185-1332 185-1334 185-1341 185-1342 185-1344 185-1346
Spare seal rings for small flange connections
Forsegling Tætningsringe
These spare seal rings for small flange KF connections are made of NBR or FKM.