Capillary columns, Elite-FFAP for autosystem
Leverandør: PerkinElmer
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
N9316351 N9316352 N9316353 N9316354 N9316357 N9316358
PERCN9316351EA 7830 DKK
PERCN9316351 PERCN9316352 PERCN9316353 PERCN9316354 PERCN9316357 PERCN9316358
Capillary columns, Elite-FFAP for autosystem
Kromatografikolonner
Elite-FFAP columns consist of nitroterephthalic acid modified polyethylene glycol (bonded).