Cell culture flasks, screw cap with 2 ports, Corning®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
11440 431514 431512 431516 431518 431520
CORN11440EA 4130 DKK
CORN11440 CORN431514 CORN431512 CORN431516 CORN431518 CORN431520
Cell culture flasks, screw cap with 2 ports, Corning®
Kolber Celledyrkningskolber
Erlenmeyer flasks made of clear polycarbonate, with PP screw cap, are now available as closed system solutions.
  • Sterilised by gamma radiation
  • Certified nonpyrogenic
They are sterile and arrive individually double-bagged and peassembled. The preassembled transfer caps have two ports. One port ends in a 0,2 µm filter, and the other port is chemically resistant, heat sealable flexible tubing, ending in a male Luer lock. The dip tube reaches all the way to the bottom of the flask for easy aseptic transfer of your liquid handling processes.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904.