2-Benzylcyclohexanone ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

L05776.03 L05776.06
L05776.03EA 558 DKK
L05776.03 L05776.06
2-Benzylcyclohexanone ≥97%
2-Benzylcyclohexanone

Purity: 97% CAS No.: 946-33-8 Molecular Formula: C13H16O Formula Weight: 188.27 Flash Point: 110°(230°F) Refractive Index: 1.5360 Melting Point: 28-30° Boiling Point: 103-105°/0.2mm Beilstein Reference: 2046670 MDL No.: MFCD00051455

Formel: C₁₃H₁₆O
Molekylvægt: 188.27 g/mol
Kogepunkt: 103…105 °C (0.2 mmHg)
Smeltepunkt: 28…30 °C
Massefylde: 1.030
Flammepunkt: >110 °C (230 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00051455
CAS-nummer: 946-33-8
EINECS: 213-420-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR