Ethyl thiophene-2-glyoxylate ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Ethyl 2-oxo-2-(thiophen-2-yl)acetate

B24712.09 B24712.18
B24712.09EA 517 DKK
B24712.09 B24712.18
Ethyl thiophene-2-glyoxylate ≥97%
Ethyl thiophene-2-glyoxylate

Purity: 97% CAS No.: 4075-58-5 Molecular Formula: C8H8O3S Formula Weight: 184.21 Flash Point: 110°(230°F) Refractive Index: 1.5505 Boiling Point: 102-110°/0.8mm Beilstein Reference: 5651 MDL No.: MFCD00015538

Formel: C₈H₈O₃S
Kogepunkt: 102…110 °C (0,8 mmHg)
Massefylde: 1,250
Flammepunkt: >110 °C (230 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00015538
CAS-nummer: 4075-58-5
EINECS: 223-793-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR