Tetrabromobisphenol A 97

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4,4'-(Propane-2,2-diyl)bis(2,6-dibromophenol), 3,3',5,5'-Tetrabromobisphenol A

A17395.22 A17395.36
A17395.22EA 623 DKK
A17395.22 A17395.36
Tetrabromobisphenol A 97
Tetrabromobisphenol A

Purity: 97% CAS No.: 79-94-7 Molecular Formula: C15H12Br4O2 Formula Weight: 543.87 Odour: Odourless Melting Point: 179-182° UN No.: 3077 MDL No.: MFCD00013962

Formel: C₁₅H₁₂Br₄O₂
Molekylvægt: 543.87 g/mol
Smeltepunkt: 179…182 °C
Massefylde: 1.76
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00013962
CAS-nummer: 79-94-7
EINECS: 201-236-9
ADR: 9 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR