2-Phenylmalonamide ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B24972.03 B24972.06 B24972.14
B24972.03EA 205 DKK
B24972.03 B24972.06 B24972.14
2-Phenylmalonamide ≥97%
2-Phenylmalonamide

Purity: 97% CAS No.: 10255-95-5 Molecular Formula: C9H10N2O2 Formula Weight: 178.19 Melting Point: ca 233° dec. Beilstein Reference: 2446518

Formel: C₉H₁₀N₂O₂
Molekylvægt: 178.19 g/mol
Smeltepunkt: ∼ 233 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00051776
CAS-nummer: 10255-95-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR