Acetohydroxamic acid 98+%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
N-Hydroxyacetamide
170130050
170130250
ACRO170130050EA
610
DKK
ACRO170130050
ACRO170130250
Acetohydroxamic acid 98+%
Acetohydroxamic acid
Formel:
C₂H₅NO₂ Smeltepunkt: 86…90 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00009994 CAS-nummer: 546-88-3 EINECS: 208-913-8 |
Specifikationer Testresultater
Appearance | White to light yellow Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 85 to 90 °C |
GC | ≥98.0 % |
Titration with NaOH | 98.0 to 102.0 % |
Specific optical rotation | -24° to -10° (20 °C, 589 nm) (c=1,H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...