Carminic acid 70-90%
Leverandør: Thermo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
ACRO346630050EA
2600
DKK
ACRO346630050
ACRO346630010
Carminic acid 70-90%
Carminic acid
Formel:
C₂₂H₂₀O₁₃ Smeltepunkt: 136 °C (dec.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00167028 CAS-nummer: 1260-17-9 EINECS: 215-023-3 UN nummer: 3147 ADR: 8,III |
Specifikationer Testresultater
Appearance | Red Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥70 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...