4-Methoxybenzhydrol ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
L00468.06
L00468.14
L00468.06EA
401
DKK
L00468.06
L00468.14
4-Methoxybenzhydrol ≥98%
4-Methoxybenzhydrol
Purity: 98+%
CAS No.: 720-44-5
Molecular Formula: C14H14O2
Formula Weight: 214.26
Storage Temperature: 2°C to 8°C
Melting Point: 65-68°
Beilstein Reference: 3202445
MDL No.: MFCD00014398
Formel:
C₁₄H₁₄O₂ Molekylvægt: 214.26 g/mol Smeltepunkt: 65…68 °C |
MDL Number:
MFCD00014398 CAS-nummer: 720-44-5 EINECS: 211-953-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...