4-Fluor-3-trifluormethylphenol ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-Fluoro-5-hydroxybenzotrifluoride, 4-Fluoro-3-trifluoromethylphenol

F0754-25G F0754-5G
TCIAF0754-25GEA 1330 DKK
TCIAF0754-25G TCIAF0754-5G
4-Fluor-3-trifluormethylphenol ≥98.0% (by GC, titration analysis)
4-Fluor-3-trifluormethylphenol
Formel: C₇H₄F₄O
Molekylvægt: 180.1 g/mol
Kogepunkt: 86°C
Flammepunkt: 79°C
MDL Number: MFCD00043875
CAS-nummer: 61721-07-1
EINECS: 432-560-0
UN nummer: 3265
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c62b6cd800229603d5d19eedcd6a52ffa01e6b7&vid=2c62b6cd800229603d5d19eedcd6a52ffa01e6b7&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR