Super syringes
Leverandør: HAMILTON BONADUZ
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
86311 86302 86312 86313 86314 86301
HAMI86311EA 10520 DKK
HAMI86311 HAMI86302 HAMI86312 HAMI86313 HAMI86314 HAMI86301
Super syringes
Sprøjter Sprøjter til kromatografi
Super syringes with PTFE Luer lock (TLL) connector or tracheal adapter.
  • Clear acrylic syringe barrels
  • For use with gases and liquids
Ideal for use with air sampling, preparing gas standards, calibrating reservoirs, and pneumographs.