Syringes, FORTUNA® OPTIMA®
Leverandør: Poulten & Graf
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7.20227 7.20233 7.20241 7.24027 7.24033 7.24037 7.24041 7.24044 7.20237 7.20244
612-0382EA 2330 DKK
612-0382 612-0383 612-0385 612-0392 612-0393 612-0394 612-0395 613-0495 612-0384 POUL7.20244
Syringes, FORTUNA® OPTIMA®
Sprøjter Sprøjter, medicinske
These syringes are made of Durobax® glass with steel piston.
  • Interchangeable
  • Amber graduation