Blandingsmålecylindre, høj form, klasse A, USP, BLAUBRAND®
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USP32408 USP32420 USP32428 USP32438 USP32448 USP32454 USP32462
BRNDUSP32408EA 710 DKK
BRNDUSP32408 BRNDUSP32420 BRNDUSP32428 BRNDUSP32438 BRNDUSP32448 BRNDUSP32454 BRNDUSP32462
Blandingsmålecylindre, høj form, klasse A, USP, BLAUBRAND®
Cylindere Målecylindre
Borosilicate glass 3.3, class A, conformity certified.
  • With individual USP certificate
  • Calibrated to contain (TC, In)
  • Hexagonal glass base, PP stopper (except 500 and 1000 ml capacity cylinders, PE stopper)
  • Ring marks at major graduations, marks and inscriptions in high contrast blue enamel
* Height without stopper.
Certificeringer: DIN EN ISO 4788.