Målecylindre, høj form, klasse A, borosilikatglas, DAkkS-certifikat, BLAUBRAND® ETERNA
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
DAKKS32762 DAKKS32720 DAKKS32738 DAKKS32754 DAKKS32764 DAKKS32708
BRNDDAKKS32762EA 1560 DKK
BRNDDAKKS32762 BRNDDAKKS32720 BRNDDAKKS32738 BRNDDAKKS32754 BRNDDAKKS32764 BRNDDAKKS32708
Målecylindre, høj form, klasse A, borosilikatglas, DAkkS-certifikat, BLAUBRAND® ETERNA
Cylindere Målecylindre
Borosilicate 3.3 glass, tall form, class A, conformity certified.
  • DAkkS calibration certificate included
  • Calibrated to contain (TC, In)
  • Hexagonal glass base, with spout
  • Ring marks at major graduations, marks and inscriptions in ETERNA amber stain
Certificeringer: DIN EN ISO 4788