Burettes, straight stopcock, Schellbach, with USP individual or DAkkS calibration certificate, BLAUBRAND®
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
DAKKS12484 USP12484 USP12487 USP12488 DAKKS12488
BRNDDAKKS12484EA 1800 DKK
BRNDDAKKS12484 BRNDUSP12484 BRNDUSP12487 BRNDUSP12488 BRNDDAKKS12488
Burettes, straight stopcock, Schellbach, with USP individual or DAkkS calibration certificate, BLAUBRAND®
Buretter
Borosilicate glass 3.3, class AS.
  • Calibrated to deliver 'TD, Ex', conformity certified
  • Schellbach stripe, ring marks at major graduations
  • Marks and inscriptions in high contrast blue enamel represent the optimum combination of resistance and readability
Length: 800 mm