Laboratory bottles, narrow neck, with thread
Leverandør: witeg Labortechnik
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5.061.020 5.527.005
215-1562EA 8880 DKK
215-1562 WITG5.527.005
Laboratory bottles, narrow neck, with thread
Flasker Glasflasker
These laboratory bottles are made of borosilicate glass with safety coating witofoam.
  • Excellent chemical resistance, good heat resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Compressive strength is not increased by the plastic coating
Round, with graduation and DIN thread. The coating provides scratch, leak and splinter protection and is ideally suited to both the transport and storage of hazardous media or valuable samples.