Woulff bottles
Leverandør: witeg Labortechnik
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5.810.500 5.810.010 5.810.005 5.810.002
WITG5.810.500EA 1580 DKK
WITG5.810.500 WITG5.810.010 WITG5.810.005 WITG5.810.002
Woulff bottles
Flasker Woulff-flasker
Made of borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion gives relatively high resistance to temperature changes
With three necks.