Eppendorf® Xplorer® Plus elektroniske enkeltkanalspipetter
Leverandør: EPPENDORF
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4861000.708 4861000.716 4861000.724 4861000.732 4861000.740 4861000.759 4861000.720 4861000.736 4861000.710
613-2229EA 6440 DKK
613-2229 613-2230 613-2231 613-2232 613-2233 613-2234 613-6984 613-6985 613-6983
Eppendorf® Xplorer® Plus elektroniske enkeltkanalspipetter
Pipetter Automatiske pipetter
Eppendorf® Xplorer® plus electronic single-channel pipettes provide an accurate, precise, ergonomic liquid handling solution, featuring an intuitive interface, programmable modes, and advanced charging technology for reliable, efficient pipetting, enhancing reproducibility and minimising errors.
  • Multifunctional rocker: 'Up is up and down is down™' principle
  • Clearly arranged color display; no sub-menus; help function
  • Innovative rocker: When the tip is ejected, the piston automatically returns to zero position
  • ON/OFF switch: Switches the pipetter off during longer periods of inactivity
  • Programming: One-time programming of sequences for easier handling of complex processes
  • Individual fixed volume setting: Desired volume can be saved
  • Password protection: Protects programming and adjustment
  • Individual speed setting: Sets parameters to your individual working speed
Reliably reproducible results:
Tips always rest in the same position
Volume and speed precisely and reproducibly adjustable
Adjustable to your work environment and requirements
PhysioCare Concept™:
Reduced weight
Improved ergonomics
Perfect balance
Optimally angled pipetter head
Improved view and high resolution display
Spring-loaded tip cone provides maximum tightness
Easy selection of all functions with the practical selection dial. Air-cushion pipetting system, with autoclavable lower part and epT.I.P.S.® consumables. It operates in the language of your choice. Pipetter maintenance reminder function, based on time or frequency of use as desired.
Applications: Pipetting liquids (manual and automatic), dispensing (timed and manual), pipetting and mixing. Additional applications include aspirating, diluting, sequential dispensing and reverse pipetting.
Bestillingsoplysninger: Supplied with charger.