Anti-static harnesses, Miller® ATEX
Leverandør: HONEYWELL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1015074 1015075
113-1275EA 3340 DKK
113-1275 113-1276
Anti-static harnesses, Miller® ATEX
Faldbeskyttelse Faldseler
This harness is the perfect solution for work at height in explosive atmospheres. It is particularly suitable for petrochemical and mining environments. The anti-static material prevents the risks of an electrostatic discharge igniting the explosive atmosphere thus allowing the user to work in complete safety in these high-risk environments.
  • With two anchorage points (rear and front): Versatile
  • Shoulder and leg straps made of 45 mm polyester webbing (anti-static) with steel threads in the webbing to ensure total conductibility
  • Dorsal extension strap facilitates connection to the intermediate attachment (lanyard or fall arrest block)
  • Detachable dorsal pad facilitates harness donning and prevents the straps from tangling up
Certificeringer: EN 361, ATEX 949/CE, EN 13464-1, EN 13463-1