Labelling tapes, industrial grade vinyl tape
Leverandør: MICRONOVA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
47B-12WH 47B-34WH 47B-3WH 47B-14BL 47B-12BL 47B-12TR 47B-12DB 47B-12RE 47B-2OR 47B-12OR 47B-2LG 47B-12DG 47B-12YE 47B-1TR 47B-1YE 47B-12WHIRM 47B-1DB 47B-1RE 47B-1DG 47B-15WH 47B-2DB 47B-2DG 47B-2TR 47B-2YE
817-0226EA 124 DKK
817-0226 817-0254 817-0260 817-0228 817-0219 817-0225 817-0221 817-0224 817-0244 817-0223 817-0243 817-0222 817-0227 817-0236 817-0237 817-0487 817-0488 817-0490 817-0489 MICR47B-15WH MICR47B-2DB MICR47B-2DG MICR47B-2TR MICR47B-2YE
Labelling tapes, industrial grade vinyl tape
Tape
Processed in a controlled environment, tapes are wound on a plastic core and single bagged. Ideal for wheel taping and colour coding applications such as spray bottle identification.
  • Forms strong bond without leaving adhesive residue
  • Can be written on with cleanroom pen
  • Different colours available
Length: 32,9 m