Phthalhydrazide 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

131061000 131060250
ACRO131061000EA 742 DKK
ACRO131061000 ACRO131060250
Phthalhydrazide 99%
Phthalhydrazide
Formel: C₈H₆N₂O₂
Smeltepunkt: >300 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006888
CAS-nummer: 1445-69-8
EINECS: 215-893-4

Bestil nu

Specifikationer Testresultater

Appearance White Powder and/or chunks
Infrared spectrum Conforms
Loss on drying ≤1 %
Titration with NaOH 98.5 to 101.5 % (On dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR