Diphenylboric acid-2-aminoethyl ester (2-APB)
Leverandør: Cayman Chemical
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: 2-Aminoethyl diphenylborinate, 2-((Diphenylboryl)oxy)ethanamine
64970-1 64970-5
CAYM64970-1EA 678 DKK
CAYM64970-1 CAYM64970-5
Diphenylboric acid-2-aminoethyl ester (2-APB)
Diphenylboric acid-2-aminoethyl ester (2-APB)
Formel: C₁₄H₁₆BNO
Molekylvægt: 225.1 g/mol
MDL Number: MFCD00014823
CAS-nummer: 524-95-8
EINECS: 208-366-5