Dibenzyl diisopropylphosphoramidite 94%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B22629.06
B22629.14
B22629.22
B22629.PF
B22629.06EA
1120
DKK
B22629.06
B22629.14
B22629.22
B22629.PF
Dibenzyl diisopropylphosphoramidite 94%
Dibenzyl diisopropylphosphoramidite
Purity: 95%
CAS No.: 108549-23-1
Molecular Formula: C20H28NO2P
Formula Weight: 345.43
Storage Temperature: 2°C to 8°C
Flash Point: 69°(156°F)
Physical Form: Liquid
Density: 1.030
Sensitivity: Moisture Sensitive
Boiling Point: 130°/0.55mm
MDL No.: MFCD00191988
Formel:
C₂₀H₂₈NO₂P Kogepunkt: 130 °C (0,55 mmHg) Massefylde: 1,030 Flammepunkt: 69 °C (156 °F) |
MDL Number:
MFCD00191988 CAS-nummer: 108549-23-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...