4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2)
Leverandør: Serva
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Disodium 4-methylumbelliferyl phosphate, 4-MUP
29670.01
29670.02
SERA29670.01EA
0
DKK
SERA29670.01
SERA29670.02
4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2)
4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2)
Formel:
C₁₀H₇Na₂O₆P |
MDL Number:
MFCD00037614 CAS-nummer: 22919-26-2 EINECS: 245-325-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...