5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate)
Leverandør: Serva
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: BCIP, P-toluidine 5-bromo-4-chloro-1H-indol-3-yl phosphate
15247.02 15247.03
SERA15247.02EA 0 DKK
SERA15247.02 SERA15247.03
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate)
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate)
Formel: C₁₅H₁₅BrClN₂O₄P
Molekylvægt: 433.6214 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD00040636
CAS-nummer: 6578-06-9
EINECS: 229-506-1