N,N-Diethylhydroxylamine 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B23302.14 B23302.22
B23302.14EA 509 DKK
B23302.14 B23302.22
N,N-Diethylhydroxylamine 97%
N,N-Diethylhydroxylamine

Purity: 97% CAS No.: 3710-84-7 Molecular Formula: C4H11NO Formula Weight: 89.14 Flash Point: 46°(114°F) Refractive Index: 1.4215 Melting Point: -25° Boiling Point: 125-130° Beilstein Reference: 1731349 UN No.: 1993 MDL No.: MFCD00002126

Formel: C₄H₁₁NO
Molekylvægt: 89.14 g/mol
Kogepunkt: 125…130 °C
Smeltepunkt: –25 °C
Massefylde: 0.867
Flammepunkt: 46 °C (114 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002126
CAS-nummer: 3710-84-7
EINECS: 223-055-4
ADR: 3 III

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR