N,N'-Diacetylhydrazine ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
N'-Acetylacetohydrazide
A13393.09
A13393.18
A13393.30
A13393.09EA
477
DKK
A13393.09
A13393.18
A13393.30
N,N'-Diacetylhydrazine ≥98%
N,N'-Diacetylhydrazine
Purity: 98%
CAS No.: 3148-73-0
Molecular Formula: CH3CONHNHCOCH3
Formula Weight: 116.12
Melting Point: 138-142°
Boiling Point: 209°/15mm
MDL No.: MFCD00008673
Formel:
C₄H₈N₂O₂ Kogepunkt: 209 °C (15 mmHg) Smeltepunkt: 138…142 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008673 CAS-nummer: 3148-73-0 EINECS: 221-576-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...