Cyclobutanecarbonitrile ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

A10309.06 A10309.14
A10309.06EA 1500 DKK
A10309.06 A10309.14
Cyclobutanecarbonitrile ≥97%
Cyclobutanecarbonitrile

Purity: 97% CAS No.: 4426-11-3 Molecular Formula: C5H7N Formula Weight: 81.12 Flash Point: 42°(107°F) Refractive Index: 1.4325 Odour: Recognizable Boiling Point: 144-146° Beilstein Reference: 2037573 UN No.: 1992 MDL No.: MFCD00013735

Formel: C₅H₇N
Molekylvægt: 81.12 g/mol
Kogepunkt: 144…146 °C
Massefylde: 0.868
Flammepunkt: 42 °C (107 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00013735
CAS-nummer: 4426-11-3
EINECS: 224-606-1
UN nummer: 1992
ADR: 3 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR